Product Details

SNP ID
rs75714902
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:124805675 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGATAGTGAGAAGACCAAGAAGAAA[A/G]AGAATGTAAGGTAAAGGGGGCAGAG
Phenotype
MIM: 611144
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FAM175B PubMed Links

Gene Details

Gene
FAM175B
Gene Name
family with sequence similarity 175 member B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032182.3 Intron NP_115558.3
XM_011539554.1 Intron XP_011537856.1
XM_017015986.1 Intron XP_016871475.1

View Full Product Details