Product Details

SNP ID
rs74694974
Assay Type
Functionally tested
NCBI dbSNP Submissions
10
Location
Chr.1:40377374 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTACTTCGTACAAGTTTTGTTGAT[C/T]CTGGGGTTTTTAACTGTCCTGGGCA
Phenotype
MIM: 616916
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SMAP2 PubMed Links

Gene Details

Gene
SMAP2
Gene Name
small ArfGAP2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001198978.1 Intron NP_001185907.1
NM_001198979.1 Intron NP_001185908.1
NM_001198980.1 Intron NP_001185909.1
NM_022733.2 Intron NP_073570.1
XM_011541960.1 Intron XP_011540262.1
XM_017002093.1 Intron XP_016857582.1

View Full Product Details