Product Details

SNP ID
rs76715802
Assay Type
Functionally tested
NCBI dbSNP Submissions
16
Location
Chr.1:54785043 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGTGTGCTTGATGCTGAGTTTTAC[A/G]TACAAAATCTAATGTAATTCTCATA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TTC22 PubMed Links

Gene Details

Gene
TTC22
Gene Name
tetratricopeptide repeat domain 22
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001114108.1 1727 Intron NP_001107580.1
NM_017904.3 1727 UTR 3 NP_060374.2
XM_011541671.2 1727 Intron XP_011539973.1
XM_017001582.1 1727 Intron XP_016857071.1

View Full Product Details