Product Details

SNP ID
rs75425953
Assay Type
Functionally tested
NCBI dbSNP Submissions
14
Location
Chr.1:32890296 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGTACGCTCATACTGACTTCACAA[A/G]GAGATTTAAGGACCGGATATAAATG
Phenotype
MIM: 142622
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
HPCA PubMed Links

Gene Details

Gene
HPCA
Gene Name
hippocalcin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002143.2 Intron NP_002134.2
XM_005270792.2 Intron XP_005270849.1
XM_017001118.1 Intron XP_016856607.1
Gene
TMEM54
Gene Name
transmembrane protein 54
There are no transcripts associated with this gene.

View Full Product Details