Product Details

SNP ID
rs80304058
Assay Type
Functionally tested
NCBI dbSNP Submissions
13
Location
Chr.1:33272112 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGAGAGGCTAGGGACTGTAAAGGAG[C/G]CTGAGAAAGTGAGGAGCCAGGTGTC
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ZNF362 PubMed Links

Gene Details

Gene
ZNF362
Gene Name
zinc finger protein 362
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152493.2 Intron NP_689706.2
XM_005270507.2 Intron XP_005270564.1
XM_005270508.4 Intron XP_005270565.1
XM_017000415.1 Intron XP_016855904.1

View Full Product Details