Product Details

SNP ID
rs76601224
Assay Type
Functionally tested
NCBI dbSNP Submissions
7
Location
Chr.1:33478354 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACACAGCGTGGTGATGAATTGGATT[G/T]GGGGATCTTAGAGAATGATGATGAC
Phenotype
MIM: 611315
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
ZSCAN20 PubMed Links

Gene Details

Gene
ZSCAN20
Gene Name
zinc finger and SCAN domain containing 20
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_145238.3 Intron NP_660281.2
XM_005271171.3 Intron XP_005271228.1
XM_006710874.3 Intron XP_006710937.1
XM_006710876.3 Intron XP_006710939.1
XM_017002237.1 Intron XP_016857726.1
XM_017002238.1 Intron XP_016857727.1
XM_017002239.1 Intron XP_016857728.1
XM_017002240.1 Intron XP_016857729.1
XM_017002241.1 Intron XP_016857730.1

View Full Product Details