Product Details

SNP ID
rs76988303
Assay Type
Functionally tested
NCBI dbSNP Submissions
13
Location
Chr.1:74706455 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGCTGCTGCATATTGCTGAAATTC[C/T]TCCTAAGAAAAGGAAAAACAAATTT
Phenotype
MIM: 123691
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CRYZ PubMed Links

Gene Details

Gene
CRYZ
Gene Name
crystallin zeta
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001130042.1 820 Silent Mutation GAA,GAG E277E NP_001123514.1
NM_001130043.1 820 Silent Mutation GAA,GAG E243E NP_001123515.1
NM_001134759.1 820 Silent Mutation GAA,GAG E140E NP_001128231.1
NM_001889.3 820 Silent Mutation GAA,GAG E277E NP_001880.2
XM_005270491.4 820 Silent Mutation GAA,GAG E140E XP_005270548.1
XM_011540747.1 820 Silent Mutation GAA,GAG E277E XP_011539049.1
XM_017000367.1 820 Silent Mutation GAA,GAG E243E XP_016855856.1
XM_017000368.1 820 Silent Mutation GAA,GAG E140E XP_016855857.1

View Full Product Details