Product Details
- SNP ID
-
rs78397609
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
13
- Location
-
Chr.1:78235291 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GTAGTGTAGTGGAAAGACCATGGTA[C/T]TTTGGACCATGCAGTAAGAACATGG
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
MGC27382
PubMed Links
Gene Details
- Gene
- MGC27382
- Gene Name
- uncharacterized MGC27382
There are no transcripts associated with this gene.
View Full Product Details