Product Details

SNP ID
rs78414260
Assay Type
Functionally tested
NCBI dbSNP Submissions
11
Location
Chr.1:85255713 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATAAAAAAGTCACTTGCTAATCTA[A/C]AATCTGGGTAAGAGTGTAGAGTTTC
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
C1orf52 PubMed Links

Gene Details

Gene
C1orf52
Gene Name
chromosome 1 open reading frame 52
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_198077.3 Intron NP_932343.1

View Full Product Details