Product Details

SNP ID
rs78294528
Assay Type
Functionally tested
NCBI dbSNP Submissions
10
Location
Chr.1:35183225 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AACTAAACCTACTTCAGTACTAAAC[C/T]TTTTTTTTTTTTTGAGACAGAGTCT
Phenotype
MIM: 605199
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SFPQ PubMed Links

Gene Details

Gene
SFPQ
Gene Name
splicing factor proline and glutamine rich
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005066.2 3467 Intron NP_005057.1
XM_005271112.4 3467 UTR 3 XP_005271169.1
XM_005271113.4 3467 UTR 3 XP_005271170.1
XM_005271115.4 3467 Intron XP_005271172.1
XM_011541950.2 3467 UTR 3 XP_011540252.1
XM_017002053.1 3467 UTR 3 XP_016857542.1
XM_017002054.1 3467 UTR 3 XP_016857543.1

View Full Product Details