Product Details
- SNP ID
-
rs76666228
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
12
- Location
-
Chr.1:92037558 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CTGAGGCAGGCAATTTCTTTGAGTC[A/G]TCTTCGTCTCCAGCTATATGGAATG
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
EPHX4
PubMed Links
Gene Details
- Gene
- EPHX4
- Gene Name
- epoxide hydrolase 4
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_173567.4 |
|
Intron |
|
|
NP_775838.3 |
View Full Product Details