Product Details

SNP ID
rs75271503
Assay Type
Functionally tested
NCBI dbSNP Submissions
19
Location
Chr.1:35870694 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TATTATTTTTACCCTTATTTTGGTT[G/T]TTTACCTTTATCTTTTGAGATATTC
Phenotype
MIM: 606228
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
AGO1 PubMed Links

Gene Details

Gene
AGO1
Gene Name
argonaute 1, RISC catalytic component
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001317122.1 Intron NP_001304051.1
NM_001317123.1 Intron NP_001304052.1
NM_012199.4 Intron NP_036331.1
XM_011541236.2 Intron XP_011539538.1
XM_017001016.1 Intron XP_016856505.1
XM_017001017.1 Intron XP_016856506.1
XM_017001018.1 Intron XP_016856507.1
XM_017001019.1 Intron XP_016856508.1

View Full Product Details