Product Details

SNP ID
rs77457406
Assay Type
Functionally tested
NCBI dbSNP Submissions
13
Location
Chr.1:113878068 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGCATCTGTGAGAGACAGGGTTAA[C/T]GGAGAGAGAAAAAAGTCAAGCTAGA
Phenotype
MIM: 600716
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
AP4B1-AS1 PubMed Links

Gene Details

Gene
AP4B1-AS1
Gene Name
AP4B1 antisense RNA 1
There are no transcripts associated with this gene.

Gene
BCL2L15
Gene Name
BCL2 like 15
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001010922.2 3719 UTR 3 NP_001010922.1
Gene
PTPN22
Gene Name
protein tyrosine phosphatase, non-receptor type 22
There are no transcripts associated with this gene.

View Full Product Details