Product Details

SNP ID
rs76674123
Assay Type
Functionally tested
NCBI dbSNP Submissions
19
Location
Chr.1:116912499 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGCTCTTTTTGACCTCCATATTCA[C/T]GTAGTCTCTCAATTCTTGGCTCTCC
Phenotype
MIM: 601204
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PTGFRN PubMed Links

Gene Details

Gene
PTGFRN
Gene Name
prostaglandin F2 receptor inhibitor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020440.3 Intron NP_065173.2
XM_017001874.1 Intron XP_016857363.1

View Full Product Details