Product Details

SNP ID
rs75715616
Assay Type
Functionally tested
NCBI dbSNP Submissions
16
Location
Chr.1:117610377 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTCTCCCTAACTGCCCTTCTTTGC[C/T]TCACAGAACAGATACTCATATCTGC
Phenotype
MIM: 613952
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FAM46C PubMed Links

Gene Details

Gene
FAM46C
Gene Name
family with sequence similarity 46 member C
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017709.3 Intron NP_060179.2
Gene
LOC100996263
Gene Name
uncharacterized LOC100996263
There are no transcripts associated with this gene.

View Full Product Details