Product Details

SNP ID
rs79962437
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:114062810 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCTGGCGTGAAGCTGACAAGGAAG[A/G]GGCTAAAGTCTTGCTAATTTTAAGT
Phenotype
MIM: 176797
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZBTB16 PubMed Links

Gene Details

Gene
ZBTB16
Gene Name
zinc finger and BTB domain containing 16
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001018011.1 591 Intron NP_001018011.1
NM_006006.4 591 Intron NP_005997.2
XM_005271658.4 591 Intron XP_005271715.1
XM_017018257.1 591 Intron XP_016873746.1
XM_017018258.1 591 Intron XP_016873747.1
XM_017018259.1 591 UTR 5 XP_016873748.1

View Full Product Details