Product Details

SNP ID
rs77872571
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:130405477 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCTGCCTTGATATGGCCAAGGGAC[C/T]CAGTCACCACAGTGGAGACCCTTGT
Phenotype
MIM: 605175
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ADAMTS8 PubMed Links

Gene Details

Gene
ADAMTS8
Gene Name
ADAM metallopeptidase with thrombospondin type 1 motif 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_007037.5 3458 UTR 3 NP_008968.4
XM_017017145.1 3458 UTR 3 XP_016872634.1
Gene
LOC646383
Gene Name
uncharacterized LOC646383
There are no transcripts associated with this gene.

View Full Product Details