Product Details

SNP ID
rs79317118
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:55903920 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GATTCTTTATGGCTTTCTACTCCAA[C/T]CTAAGAGTGGCAGGATGCCCTGAAT
Phenotype
MIM: 616181
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZNF713 PubMed Links

Gene Details

Gene
ZNF713
Gene Name
zinc finger protein 713
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_182633.2 Intron NP_872439.2

View Full Product Details