Product Details

SNP ID
rs74616725
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:95655498 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTCACCCCAACCCCTTAGACCTACT[A/G]CATTTAAACAGAGAGACATGCATTG
Phenotype
MIM: 608039
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
RFESD PubMed Links

Gene Details

Gene
RFESD
Gene Name
Rieske Fe-S domain containing
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001131065.1 Intron NP_001124537.1
NM_001131066.1 Intron NP_001124538.1
NM_173362.3 Intron NP_775498.1
XM_011543360.2 Intron XP_011541662.1
XM_011543361.2 Intron XP_011541663.1
XM_017009413.1 Intron XP_016864902.1
Gene
SPATA9
Gene Name
spermatogenesis associated 9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_031952.3 Intron NP_114158.2
XM_011543664.1 Intron XP_011541966.1
XM_011543666.1 Intron XP_011541968.1
XM_011543667.2 Intron XP_011541969.1
XM_017009948.1 Intron XP_016865437.1
XM_017009949.1 Intron XP_016865438.1
XM_017009950.1 Intron XP_016865439.1
XM_017009951.1 Intron XP_016865440.1

View Full Product Details