Product Details

SNP ID
rs79988174
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:126360426 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCTGGAAGAGTATGTTCCACGGGA[C/G]AGAAGTGAAGCCAGTGGGTCCGGAC
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
GRAMD3 PubMed Links

Gene Details

Gene
GRAMD3
Gene Name
GRAM domain containing 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001146319.1 331 Missense Mutation ACA,AGA T32R NP_001139791.1
NM_001146320.1 331 Intron NP_001139792.1
NM_001146321.1 331 Intron NP_001139793.1
NM_001146322.1 331 Intron NP_001139794.1
NM_023927.2 331 Intron NP_076416.2
XM_005272057.3 331 Intron XP_005272114.1
XM_005272058.3 331 Intron XP_005272115.1
XM_005272059.4 331 Intron XP_005272116.1
XM_005272060.3 331 Intron XP_005272117.1
XM_005272061.3 331 Intron XP_005272118.1
XM_005272062.3 331 Intron XP_005272119.1
XM_005272063.3 331 Intron XP_005272120.1
XM_005272064.3 331 Intron XP_005272121.1
XM_011543591.2 331 Intron XP_011541893.1
XM_011543592.2 331 Intron XP_011541894.1
XM_011543593.2 331 Missense Mutation ACA,AGA T32R XP_011541895.1
XM_011543594.2 331 Intron XP_011541896.1
XM_011543595.2 331 Intron XP_011541897.1
XM_017009781.1 331 UTR 5 XP_016865270.1
XM_017009782.1 331 Intron XP_016865271.1
XM_017009783.1 331 Intron XP_016865272.1
XM_017009784.1 331 Intron XP_016865273.1
XM_017009785.1 331 Intron XP_016865274.1
Gene
LOC105369178
Gene Name
uncharacterized LOC105369178
There are no transcripts associated with this gene.

View Full Product Details