Product Details

SNP ID
rs74977087
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:135581604 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTAACAACACCAGCGATCGTGGGCA[C/T]TGAGTTTTCACAGGAAAAAGTAGTT
Phenotype
MIM: 604186 MIM: 616150
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CXCL14 PubMed Links

Gene Details

Gene
CXCL14
Gene Name
C-X-C motif chemokine ligand 14
There are no transcripts associated with this gene.

Gene
SLC25A48
Gene Name
solute carrier family 25 member 48
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_145282.4 Intron NP_660325.4
XM_005271903.1 Intron XP_005271960.1
XM_006714544.1 Intron XP_006714607.1
XM_006714546.1 Intron XP_006714609.1
XM_006714550.1 Intron XP_006714613.1
XM_006714552.2 Intron XP_006714615.1
XM_011543196.1 Intron XP_011541498.1
XM_011543199.1 Intron XP_011541501.1
XM_017009086.1 Intron XP_016864575.1
XM_017009087.1 Intron XP_016864576.1
XM_017009088.1 Intron XP_016864577.1
XM_017009089.1 Intron XP_016864578.1
XM_017009090.1 Intron XP_016864579.1
XM_017009091.1 Intron XP_016864580.1
XM_017009092.1 Intron XP_016864581.1

View Full Product Details