Product Details
- SNP ID
-
rs80000074
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.8:10607380 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TCACCTTGAGTTTCTCCTTCTGACT[C/G]TGGCTGGGCCTCCCCTTCAGCCTCC
- Phenotype
-
MIM: 608581
- Polymorphism
- C/G, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
RP1L1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs56382513] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- RP1L1
- Gene Name
- retinitis pigmentosa 1-like 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_178857.5 |
6947 |
Missense Mutation |
CAG,GAG |
Q2240E |
NP_849188.4 |
View Full Product Details