Product Details
- SNP ID
-
rs77677767
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
19
- Location
-
Chr.1:229449164 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GAGCTTTGCAAAGGATTTCCAGTTT[C/T]AGATCATTTATATTTATATCTTCTT
- Phenotype
-
MIM: 607613
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
NUP133
PubMed Links
Gene Details
- Gene
- NUP133
- Gene Name
- nucleoporin 133
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_018230.2 |
3299 |
Silent Mutation |
CTA,CTG |
L1069L |
NP_060700.2 |
View Full Product Details