Product Details

SNP ID
rs79244900
Assay Type
Functionally tested
NCBI dbSNP Submissions
28
Location
Chr.1:233905571 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGGACGTCAGCCCCCGGAGACTGTC[C/T]GACATCAGCCCCCAGCTCCGGCAGC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SLC35F3 PubMed Links

Gene Details

Gene
SLC35F3
Gene Name
solute carrier family 35 member F3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001300845.1 462 Intron NP_001287774.1
NM_173508.3 462 Silent Mutation TCC,TCT S32S NP_775779.1

View Full Product Details