Product Details

SNP ID
rs79475560
Assay Type
Functionally tested
NCBI dbSNP Submissions
12
Location
Chr.1:234629337 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTGACACAAACTGCCACCTATCTC[C/G]CAAGGTTCTTTCTTTGTAACAGAAC
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
LINC00184 PubMed Links

Gene Details

Gene
LINC00184
Gene Name
long intergenic non-protein coding RNA 184
There are no transcripts associated with this gene.

View Full Product Details