Product Details

SNP ID
rs75012801
Assay Type
Functionally tested
NCBI dbSNP Submissions
14
Location
Chr.1:209787941 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGTGCTTGAGGAAACTTTAACAAA[A/C]CCAAGGAAAAATGTGGTACAAGAAC
Phenotype
MIM: 607199 MIM: 608255
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
C1orf74 PubMed Links

Gene Details

Gene
C1orf74
Gene Name
chromosome 1 open reading frame 74
There are no transcripts associated with this gene.

Gene
IRF6
Gene Name
interferon regulatory factor 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001206696.1 1938 UTR 3 NP_001193625.1
NM_006147.3 1938 UTR 3 NP_006138.1
Gene
TRAF3IP3
Gene Name
TRAF3 interacting protein 3
There are no transcripts associated with this gene.

View Full Product Details