Product Details

SNP ID
rs75522952
Assay Type
Functionally tested
NCBI dbSNP Submissions
13
Location
Chr.1:207105624 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTGGGGAAAGTGTGGGAGCCAGTT[A/C]TTTCTGGAGTTTGTAAGTGATATAA
Phenotype
MIM: 120830 MIM: 120831
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
C4BPA PubMed Links

Gene Details

Gene
C4BPA
Gene Name
complement component 4 binding protein alpha
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000715.3 Intron NP_000706.1
XM_005273251.1 Intron XP_005273308.1
XM_005273252.4 Intron XP_005273309.1
Gene
C4BPB
Gene Name
complement component 4 binding protein beta
There are no transcripts associated with this gene.

View Full Product Details