Product Details

SNP ID
rs80123208
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:27737088 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAAGATGGTAGAGAAATACGATATG[G/T]GTAGAAAACATTGGTTCCTGTGAAG
Phenotype
MIM: 602728
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
CCDC25 PubMed Links

Gene Details

Gene
CCDC25
Gene Name
coiled-coil domain containing 25
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001304529.1 Intron NP_001291458.1
NM_001304530.1 Intron NP_001291459.1
NM_001304532.1 Intron NP_001291461.1
NM_018246.2 Intron NP_060716.2
XM_011544570.2 Intron XP_011542872.1
XM_011544571.2 Intron XP_011542873.1
Gene
SCARA3
Gene Name
scavenger receptor class A member 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016240.2 Intron NP_057324.2
NM_182826.1 Intron NP_878185.1

View Full Product Details