Product Details

SNP ID
rs75301276
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:56176722 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTTTGTGGTGTTCCTGGTGATTTA[C/T]GCCATTACCTTGTTGAGGAATCTGG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
OR5J2 PubMed Links

Gene Details

Gene
OR5J2
Gene Name
olfactory receptor family 5 subfamily J member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005492.1 105 Silent Mutation TAC,TAT Y35Y NP_001005492.1

View Full Product Details