Product Details

SNP ID
rs77452976
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:68756263 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCAAGTGGTCAAAAAAATAAATACA[C/G]AATTTAAAACAATGGACAGCTAAGC
Phenotype
MIM: 600528 MIM: 604374
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CPT1A PubMed Links

Gene Details

Gene
CPT1A
Gene Name
carnitine palmitoyltransferase 1A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001031847.2 3873 Intron NP_001027017.1
NM_001876.3 3873 UTR 3 NP_001867.2
XM_005273762.2 3873 UTR 3 XP_005273819.1
XM_005273763.1 3873 Intron XP_005273820.1
XM_017017220.1 3873 Intron XP_016872709.1
Gene
TESMIN
Gene Name
testis expressed metallothionein like protein
There are no transcripts associated with this gene.

View Full Product Details