Product Details

SNP ID
rs76446931
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:74174397 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATCTAGGATTCAGTCTTTTTCTTC[A/G]GTCGTAATTTATGAAGTCTCACACC
Phenotype
MIM: 615944 MIM: 611117
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C2CD3 PubMed Links

Gene Details

Gene
C2CD3
Gene Name
C2 calcium dependent domain containing 3
There are no transcripts associated with this gene.

Gene
PPME1
Gene Name
protein phosphatase methylesterase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001271593.1 Intron NP_001258522.1
NM_016147.2 Intron NP_057231.1
XM_017017913.1 Intron XP_016873402.1

View Full Product Details