Product Details

SNP ID
rs74796152
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:74491214 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCTCTTGTCAGCAACCATAACCAA[C/T]CACGCAAGTGAATGCACCATATGGA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LIPT2 PubMed Links

Gene Details

Gene
LIPT2
Gene Name
lipoyl(octanoyl) transferase 2 (putative)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_011545021.2 1924 UTR 3 XP_011543323.2
Gene
LOC100287896
Gene Name
uncharacterized LOC100287896
There are no transcripts associated with this gene.

View Full Product Details