Product Details

SNP ID
rs77792380
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:58507314 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGAGGCTATTTTGTCTGTGTCCAC[A/G]GACTGGCTGGAGTTGGGCTGTAAGT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
OR5B21 PubMed Links

Gene Details

Gene
OR5B21
Gene Name
olfactory receptor family 5 subfamily B member 21
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005218.1 792 Silent Mutation NP_001005218.1

View Full Product Details