Product Details

SNP ID
rs79313191
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:58585296 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATAATTAAATTAGGTCAATATTAG[A/G]TCATTTTCCTTTAAGTTTGAGTGTT
Phenotype
MIM: 605390
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LPXN PubMed Links

Gene Details

Gene
LPXN
Gene Name
leupaxin
There are no transcripts associated with this gene.

Gene
ZFP91
Gene Name
ZFP91 zinc finger protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001197051.1 Intron NP_001183980.1
NM_053023.4 Intron NP_444251.1
Gene
ZFP91-CNTF
Gene Name
ZFP91-CNTF readthrough (NMD candidate)
There are no transcripts associated with this gene.

View Full Product Details