Product Details

SNP ID
rs76313618
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:59574798 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATGTTTATCACAACTAGGAAGGAA[A/G]GGATAGGGGAAGAAATAAAAGTATT
Phenotype
MIM: 167040
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
OSBP PubMed Links

Gene Details

Gene
OSBP
Gene Name
oxysterol binding protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002556.2 4683 UTR 3 NP_002547.1

View Full Product Details