Product Details

SNP ID
rs74878556
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:61339072 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCACTCCTTCCACCTCCAGGTTTCA[C/T]AGGGAAGGGGATGCTGACTGGGGTC
Phenotype
MIM: 600045 MIM: 615844
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CYB561A3 PubMed Links

Gene Details

Gene
CYB561A3
Gene Name
cytochrome b561 family member A3
There are no transcripts associated with this gene.

Gene
DDB1
Gene Name
damage specific DNA binding protein 1
There are no transcripts associated with this gene.

Gene
TKFC
Gene Name
triokinase and FMN cyclase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015533.3 587 Missense Mutation ACA,ATA T67I NP_056348.2
XM_011544912.2 587 Missense Mutation ACA,ATA T113I XP_011543214.1
XM_017017517.1 587 Missense Mutation ACA,ATA T113I XP_016873006.1
XM_017017518.1 587 Missense Mutation ACA,ATA T113I XP_016873007.1
XM_017017519.1 587 Missense Mutation ACA,ATA T67I XP_016873008.1
XM_017017520.1 587 Missense Mutation ACA,ATA T113I XP_016873009.1
XM_017017521.1 587 Missense Mutation ACA,ATA T67I XP_016873010.1
XM_017017522.1 587 Missense Mutation ACA,ATA T67I XP_016873011.1
XM_017017523.1 587 Missense Mutation ACA,ATA T67I XP_016873012.1
XM_017017524.1 587 UTR 5 XP_016873013.1

View Full Product Details