Product Details

SNP ID
rs76958090
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:55603665 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTACAATTATGTGAGACGTGCAAG[C/T]GGAGAGAGCCTTTTTCTTCCCTTTG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
OR4C11 PubMed Links

Gene Details

Gene
OR4C11
Gene Name
olfactory receptor family 4 subfamily C member 11
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004700.2 734 Missense Mutation ACT,GCT T237A NP_001004700.2

View Full Product Details