Product Details
- SNP ID
-
rs13119049
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.4:69480847 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CAAATTCAATCCAGAAGACTGCTCG[A/T]TCAAGGGGCTTCACTGGTTGATCAT
- Phenotype
-
MIM: 600067
- Polymorphism
- A/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
UGT2B4
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs13142440] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- UGT2B4
- Gene Name
- UDP glucuronosyltransferase family 2 member B4
View Full Product Details