Product Details

SNP ID
rs117371685
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:234494569 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTTTTTCCTTCTCTTTTAATTCCA[C/T]GCAACGAGGTCAGCTTTTGCAACAA
Phenotype
MIM: 604787
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ARL4C PubMed Links

Gene Details

Gene
ARL4C
Gene Name
ADP ribosylation factor like GTPase 4C
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001282431.1 2310 UTR 3 NP_001269360.1
NM_005737.3 2310 UTR 3 NP_005728.2

View Full Product Details