Product Details

SNP ID
rs111407844
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:151248937 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATCCAAGCCTCAAGGTCCAAACCTC[A/G]CAGCTGAGGTCAACAGGGGGAAAAT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
RBM43 PubMed Links

Gene Details

Gene
RBM43
Gene Name
RNA binding motif protein 43
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_198557.2 3648 UTR 3 NP_940959.1
XM_017004052.1 3648 UTR 3 XP_016859541.1

View Full Product Details