Product Details

SNP ID
rs112745900
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:151250977 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGTAAGTGTCAGAAGAAGATCCTA[C/T]AATGTCAATGTGTGTCCTATAAAGG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
RBM43 PubMed Links

Gene Details

Gene
RBM43
Gene Name
RNA binding motif protein 43
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_198557.2 1608 Missense Mutation ATA,GTA I335V NP_940959.1
XM_017004052.1 1608 Missense Mutation ATA,GTA I338V XP_016859541.1

View Full Product Details