Product Details

SNP ID
rs114975857
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:197705417 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCCGGTCTGCACCATGCTGCGAAC[A/G]TCCGTCCTCCGCCTGCTAGGACGCA
Phenotype
MIM: 609728
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MARS2 PubMed Links

Gene Details

Gene
MARS2
Gene Name
methionyl-tRNA synthetase 2, mitochondrial
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_138395.3 114 Silent Mutation ACA,ACG T4T NP_612404.1

View Full Product Details