Product Details

SNP ID
rs112181625
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:206939645 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTTTATCTTTTGGGGATGCTGGTT[C/T]CTGGAGGGCTGGGATATGATAGGTG
Phenotype
MIM: 609563
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CPO PubMed Links

Gene Details

Gene
CPO
Gene Name
carboxypeptidase O
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_173077.2 6199 Missense Mutation CCT,TCT P16S NP_775100.1
XM_017003372.1 6199 Missense Mutation CCT,TCT P16S XP_016858861.1

View Full Product Details