Product Details

SNP ID
rs114284140
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:215362410 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTTCTCTGCACTAGATGATAGAAAA[A/G]GTTTTCAAAGAATCCTAGCGATTTT
Phenotype
MIM: 601731 MIM: 135600
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ATIC PubMed Links

Gene Details

Gene
ATIC
Gene Name
5-aminoimidazole-4-carboxamide ribonucleotide formyltransferase/IMP cyclohydrolase
There are no transcripts associated with this gene.

Gene
FN1
Gene Name
fibronectin 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001306129.1 Intron NP_001293058.1
NM_001306130.1 Intron NP_001293059.1
NM_001306131.1 Intron NP_001293060.1
NM_001306132.1 Intron NP_001293061.1
NM_002026.3 Intron NP_002017.1
NM_054034.2 Intron NP_473375.2
NM_212474.2 Intron NP_997639.1
NM_212476.2 Intron NP_997641.1
NM_212478.2 Intron NP_997643.1
NM_212482.2 Intron NP_997647.1
XM_005246397.1 Intron XP_005246454.1
XM_005246398.1 Intron XP_005246455.1
XM_005246399.1 Intron XP_005246456.1
XM_005246401.1 Intron XP_005246458.1
XM_005246402.1 Intron XP_005246459.1
XM_005246403.1 Intron XP_005246460.1
XM_005246404.1 Intron XP_005246461.1
XM_005246405.1 Intron XP_005246462.1
XM_005246406.1 Intron XP_005246463.1
XM_005246407.1 Intron XP_005246464.1
XM_005246408.1 Intron XP_005246465.1
XM_005246409.1 Intron XP_005246466.1
XM_005246410.1 Intron XP_005246467.1
XM_005246411.1 Intron XP_005246468.1
XM_005246412.1 Intron XP_005246469.1
XM_005246414.1 Intron XP_005246471.1
XM_005246416.1 Intron XP_005246473.1
XM_017003692.1 Intron XP_016859181.1
XM_017003693.1 Intron XP_016859182.1
XM_017003694.1 Intron XP_016859183.1
XM_017003695.1 Intron XP_016859184.1

View Full Product Details