Product Details

SNP ID
rs112838598
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:216039318 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTAAAGGTCTCCTTCATCTTTTTG[A/G]CAACAGAAAGGTCCCCTGCTCCCTT
Phenotype
MIM: 605843
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PECR PubMed Links

Gene Details

Gene
PECR
Gene Name
peroxisomal trans-2-enoyl-CoA reductase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018441.5 944 Missense Mutation GCC,GTC A290V NP_060911.2
XM_011511478.2 944 Missense Mutation GCC,GTC A144V XP_011509780.1

View Full Product Details