Product Details

SNP ID
rs115839019
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:218881080 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCGCTCTGGGTGCTCCTGTTCTTC[C/T]TACTGCTGCTGGCTGCTGCCATGCC
Phenotype
MIM: 606268 MIM: 604663
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
WNT10A PubMed Links

Gene Details

Gene
WNT10A
Gene Name
Wnt family member 10A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_025216.2 201 Silent Mutation CTA,TTA L29L NP_079492.2
XM_011511929.2 201 Intron XP_011510231.1
XM_011511930.1 201 Silent Mutation CTA,TTA L29L XP_011510232.1
Gene
WNT6
Gene Name
Wnt family member 6
There are no transcripts associated with this gene.

View Full Product Details