Product Details

SNP ID
rs121908119
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:218882368 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATTCAGGGACCAGCGCTGGAACTG[A/C]TCAAGCCTGGAGACTCGCAACAAGA
Phenotype
MIM: 606268 MIM: 604663
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
WNT10A PubMed Links

Gene Details

Gene
WNT10A
Gene Name
Wnt family member 10A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_025216.2 437 Nonsense Mutation TGA,TGC *107C NP_079492.2
XM_011511929.2 437 Nonsense Mutation TGA,TGC *75C XP_011510231.1
XM_011511930.1 437 Nonsense Mutation TGA,TGC *107C XP_011510232.1
Gene
WNT6
Gene Name
Wnt family member 6
There are no transcripts associated with this gene.

View Full Product Details