Product Details

SNP ID
rs111741820
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:241871836 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGATGCGCATCTGGGGCCAGCGGT[A/G]CAGGCTGTGCCCCGCACCCGGGGAC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LINC01237 PubMed Links

Gene Details

Gene
LINC01237
Gene Name
long intergenic non-protein coding RNA 1237
There are no transcripts associated with this gene.

Gene
RTP5
Gene Name
receptor transporter protein 5 (putative)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_173821.2 304 Missense Mutation TAC,TGC Y94C NP_776182.2
XM_011510995.2 304 Missense Mutation TAC,TGC Y73C XP_011509297.1
XM_011510997.2 304 Missense Mutation TAC,TGC Y94C XP_011509299.1

View Full Product Details