Product Details

SNP ID
rs114762703
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:108549075 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATAGTAACGGAACTGTGTTAATGTA[C/T]ACTTTGGAGACAAACTCTCCCCAGC
Phenotype
MIM: 610643 MIM: 609929
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KIAA1524 PubMed Links

Gene Details

Gene
KIAA1524
Gene Name
KIAA1524
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020890.2 4957 Intron NP_065941.2
XM_006713716.3 4957 Intron XP_006713779.1
XM_006713717.2 4957 UTR 3 XP_006713780.1
XM_011513056.1 4957 UTR 3 XP_011511358.1
XM_011513057.2 4957 UTR 3 XP_011511359.1
Gene
MYH15
Gene Name
myosin heavy chain 15
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014981.1 4957 Intron NP_055796.1
XM_011512559.2 4957 Intron XP_011510861.1
XM_017005922.1 4957 Intron XP_016861411.1

View Full Product Details