Product Details

SNP ID
rs113125590
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:108755941 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACATGAAGCACAGACATCAGAGCTC[C/T]TGGAATTTAAGAGGAGGAAGGGCTA
Phenotype
MIM: 605645
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
RETNLB PubMed Links

Gene Details

Gene
RETNLB
Gene Name
resistin like beta
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032579.2 Intron NP_115968.1

View Full Product Details